Dimethyl-3-oxoglutarate ≥97%

Supplier: BeanTown Chemical

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

131465-100G 131465-25G 131465-500G
BT131465-100GEA 91.58 USD
BT131465-100G BT131465-25G BT131465-500G
Dimethyl-3-oxoglutarate ≥97%
Dimethyl 3-oxoglutarate

CAS: 1830-54-2; EC No: 217-385-8; MDL No: MFCD00008462 Liquid; Molecular Formula: C7H10O5; MW: 174.16 Boiling Point: 237-241°; Flash point: 110°C (230°F) Density (g/mL): 1.206; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₇H₁₀O₅
Boiling Pt: 237…241 °C
Density: 1.206 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR