tetra-Chloroauric(III) acid hydrate ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate
131445-1G
131445-5G
BT131445-1GEA
136.65
USD
BT131445-1G
BT131445-5G
tetra-Chloroauric(III) acid hydrate ≥99.999%
tetra-Chloroauric (III) acid hydrate
CAS: 27988-77-8; MDL No: MFCD00149903
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: HAuCl4·xH2O; MW: 339.79 (anhydrous)
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
HAuCl₄·H₂O |
MDL Number:
MFCD00149903 CAS Number: 27988-77-8 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...