Sodium hydrogensulfide
Supplier: BeanTown Chemical
Synonyms:
Sodium hydrogen sulfide, Sodium hydrogen sulphide
131230-1G
131230-5G
BT131230-1GEA
87.96
USD
BT131230-1G
BT131230-5G
Sodium hydrogensulfide
Sodium hydrogensulphide
CAS: 16721-80-5; EC No: 240-778-0; MDL No: MFCD00011124; RTECS: WE1900000
UN No: UN2318; Haz Class: 4.2; Packing Group: II
Crystalline; Linear Formula: NaHS; MW: 56.06
Melting Point: 350°
Density (g/mL): 1.79
Hygroscopic, Moisture Sensitive
- RTECS: WE1900000
- Hygroscopic, Moisture Sensitive
- Crystalline
Formula:
HNaS Melting Pt: 350 °C Density: 1.79 g/mL |
MDL Number:
MFCD00011124 CAS Number: 16721-80-5 EINECS: 240-778-0 UN: 2318 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...