Tetrabutyltin ≥95%
Supplier: BeanTown Chemical
Synonyms:
Tetrabutylstannane, Tetra-n-butyltin
131180-100G
131180-500G
BT131180-100GEA
56
USD
BT131180-100G
BT131180-500G
Tetrabutyltin ≥95%
Tetrabutyltin
CAS: 1461-25-2; EC No: 215-960-8; MDL No: MFCD00009422; RTECS: WH8605000
UN No: UN2788; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: [CH3(CH2)3]4Sn; Molecular Formula: C16H36Sn; MW: 347.17
Melting Point: -97°; Boiling Point: 245-247°; Flash point: 107°C (225°F)
Density (g/mL): 1.057 ; Refractive Index: 1.473
- RTECS: WH8605000
- Refractive index: 1.473
- Liquid
Formula:
C₁₆H₃₆Sn Boiling Pt: 245…247 °C Melting Pt: –97 °C Density: 1.057 g/mL Flash Pt: 107 °C (225 °F) |
MDL Number:
MFCD00009422 CAS Number: 1461-25-2 EINECS: 215-960-8 UN: 2788 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...