Diboron trioxide ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Boron oxide, Boric anhydride
131120-25G
131120-5G
BT131120-25GEA
142
USD
BT131120-25G
BT131120-5G
Diboron trioxide ≥99.999%
di-Boron trioxide
CAS: 1303-86-2; EC No: 215-125-8; MDL No: MFCD00011315; RTECS: ED7900000
Powder; Molecular Formula: B2O3 ; MW: 69.62
Melting Point: 45°
Density (g/mL): 2.46
Hygroscopic
- RTECS: ED7900000
- Hygroscopic
- Powder
Formula:
B₂O₃ Melting Pt: 45 °C Density: 2.46 g/mL |
MDL Number:
MFCD00011315 CAS Number: 1303-86-2 EINECS: 215-125-8 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...