Polyethylene oxide, M.W. > 5000000
Supplier: BeanTown Chemical
Synonyms:
PEO
131100-100G
131100-500G
BT131100-100GEA
57.15
USD
BT131100-100G
BT131100-500G
Polyethylene oxide, M.W. > 5000000
Polyethylene oxide
CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00147992
Powder; Linear Formula: (-CH2CH2O-)n
Melting Point: 175°
Density (g/mL): 1.21
Formula:
(-CH₂CH₂O-)n Melting Pt: 175 °C Density: 1.21 g/mL |
MDL Number:
MFCD00147970 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...