Lead(II) nitrate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Lead dinitrate

131000-2KG 131000-500G
BT131000-2KGEA 194.51 USD
BT131000-2KG BT131000-500G
Lead(II) nitrate ≥99.0% ACS
Lead (II) nitrate

CAS: 10099-74-8; EC No: 233-245-9; MDL No: MFCD00011153; RTECS: OG2100000 UN No: UN1469; Haz Class: 5.1 (6.1); Packing Group: II Crystalline; Linear Formula: Pb(NO3)2; MW: 331.20 Melting Point: 470° (decomposes) Density (g/mL): 4.53 Hygroscopic


  • RTECS: OG2100000
  • Hygroscopic
  • Crystalline

Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 470 °C (dec.)
Density: 4.53 g/mL
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 II
Merck Index: 13,05432

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR