HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥99%

Supplier: BeanTown Chemical

Synonyms: Hexamethyldisilazane

130970-100G 130970-25G 130970-500G
BT130970-100GEA 68.38 USD
BT130970-100G BT130970-25G BT130970-500G
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥99%
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)

CAS: 999-97-3; EC No: 213-668-5; MDL No: MFCD00008259; RTECS: JM9230000 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: (CH3)3SiNHSi(CH3)3; Molecular Formula: C6H19NSi2; MW: 161.40 Boiling Point: 125°; Flash point: 11°C (52°F) Density (g/mL): 0.774; Refractive Index: 1.408 Moisture Sensitive


  • RTECS: JM9230000
  • Refractive index: 1.408
  • Moisture Sensitive
  • Liquid

Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126
Melting Pt: -82
Density: 0.774 g/mL
Flash Pt: 11 °C (52 °F)
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 2924
ADR: 3 II
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR