Phenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

130960-10G 130960-250G 130960-50G
BT130960-10GEA 41.67 USD
BT130960-10G BT130960-250G BT130960-50G
Phenylboronic acid ≥98%
Phenylboronic acid

CAS: 98-80-6; EC No: 202-701-9; MDL No: MFCD00002103; RTECS: CY8575000 Crystalline; Molecular Formula: C6H7BO2; MW: 121.93 Melting Point: 216-219° Hygroscopic


  • RTECS: CY8575000
  • Hygroscopic
  • Crystalline

Formula: C₆H₇BO₂
MW: 121.93 g/mol
Melting Pt: 216…219 °C
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR