Molybdenum(IV) sulfide ≥99%

Supplier: BeanTown Chemical

Synonyms: Molybdenum disulfide, Molybdenum disulphide

130900-100G 130900-25G 130900-500G
BT130900-100GEA 57.08 USD
BT130900-100G BT130900-25G BT130900-500G
Molybdenum(IV) sulfide ≥99%
Molybdenum (IV) sulphide

CAS: 1317-33-5; EC No: 215-263-9; MDL No: MFCD00003470 -325 Mesh Powder; Molecular Formula: MoS2; MW: 160.08 Melting Point: 2375° Density (g/mL): 5.06


  • -325 Mesh Powder

Formula: MoS₂
Melting Pt: 2375 °C
Density: 5.06 g/mL
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR