Neodymium(III) oxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
130865-100G
130865-25G
130865-500G
BT130865-100GEA
111
USD
BT130865-100G
BT130865-25G
BT130865-500G
Neodymium(III) oxide ≥99%
Neodymium (III) oxide
CAS: 1313-97-9; EC No: 215-214-1; MDL No: MFCD00011134
Powder; Molecular Formula: Nd2O3; MW: 336.48
Melting Point: 2272°
Density (g/mL): 7.24
Hygroscopic
- Hygroscopic
- Powder
Formula:
Nd₂O₃ Melting Pt: 2272 °C Density: 7.24 g/mL |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...