Ammonium cerium(IV) nitrate ≥98.5% ACS
Supplier: BeanTown Chemical
130855-100G
130855-2KG
130855-500G
BT130855-100GEA
77.44
USD
BT130855-100G
BT130855-2KG
BT130855-500G
Ammonium cerium(IV) nitrate ≥98.5% ACS
Ammonium cerium (IV) nitrate
CAS: 16774-21-3; EC No: 240-827-6; MDL No: MFCD00151121
UN No: UN1477; Haz Class: 5.1; Packing Group: II
Granular; Linear Formula: (NH4)2Ce(NO3)6; MW: 548.23
- Granular
Formula:
(NH₄)₂Ce(NO₃)₆ |
MDL Number:
MFCD00151121 CAS Number: 16774-21-3 EINECS: 240-827-6 UN: 1477 ADR: 5.1,II |
Specification Test Results
Assay | 98.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...