3-Acetylpyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone
130795-100G
130795-25G
130795-500G
BT130795-100GEA
134
USD
BT130795-100G
BT130795-25G
BT130795-500G
3-Acetylpyridine ≥98%
3-Acetylpyridine
CAS: 350-03-8; EC No: 206-496-7; MDL No: MFCD00006396; RTECS: OB5425000
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C7H7NO; MW: 121.14
Melting Point: 11-13°; Boiling Point: 220°; Flash point: 104°C (219°F)
Density (g/mL): 1.102; Refractive Index: 1.534
- RTECS: OB5425000
- Refractive index: 1.534
- Liquid
Formula:
C₇H₇NO Boiling Pt: 220 °C Melting Pt: 11…13 °C Density: 1.102 g/mL Flash Pt: 104 °C (219 °F) |
MDL Number:
MFCD00006396 CAS Number: 350-03-8 EINECS: 206-496-7 UN: 2810 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...