2,3,6-Trifluoroaniline ≥99%
Supplier: BeanTown Chemical
130720-1G
130720-5G
BT130720-1GEA
38
USD
BT130720-1G
BT130720-5G
2,3,6-Trifluoroaniline ≥99%
2,3,6-Trifluoroaniline
CAS: 67815-56-9; MDL No: MFCD00012369
UN No: UN2941; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H4F3N; MW: 147.10
Flash point: 61°C (141°F)
Density (g/mL): 1.39
- Liquid
Formula:
C₆H₄F₃N Density: 1.39 g/mL Flash Pt: 61 °C (141 °F) |
MDL Number:
MFCD00012369 CAS Number: 67815-56-9 UN: 2941 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...