trans-Bis(tributylstannyl)ethylene ≥96%
Supplier: BeanTown Chemical
Synonyms:
trans-1,2-Bis(tri-n-butylstannyl)ethylene
130665-1G
130665-5G
BT130665-1GEA
125
USD
BT130665-1G
BT130665-5G
trans-Bis(tributylstannyl)ethylene ≥96%
trans-Bis(tributylstannyl)ethylene
CAS: 14275-61-7; MDL No: MFCD01631299
UN No: UN2788; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C26H56Sn2; MW: 606.15
Boiling Point: 185°/0.01 mmHg; Flash point:
- Refractive index: 1.502
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₂₆H₅₆Sn₂ Boiling Pt: 185 °C/0.01 mmHg Density: 1.132 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD01631299 CAS Number: 14275-61-7 ADR: 6.1,II |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...