1-Boc-4-piperidone ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

130600-10G 130600-50G
BT130600-10GEA 82 USD
BT130600-10G BT130600-50G
1-Boc-4-piperidone ≥99%
1-Boc-4-piperidone

CAS: 79099-07-3; MDL No: MFCD00151800 Powder; Molecular Formula: C10H17NO3; MW: 199.25 Melting Point: 73-77°


  • Powder

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 73…77 °C
MDL Number: MFCD00151800
CAS Number: 79099-07-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR