Scandium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

130550-25X25MM 130550-50X50MM
BT130550-25X25EA 321 USD
BT130550-25X25 BT130550-50X50
Scandium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.25 mm
Scandium

CAS: 7440-20-2; EC No: 231-129-2; MDL No: MFCD00016323 Solid; Molecular Formula: Sc; MW: 44.96 Melting Point: 1540°; Boiling Point: 2836° Density (g/mL): 2.99


  • Solid

Formula: Sc
Boiling Pt: 2836 °C
Melting Pt: 1540 °C
Density: 2.99 g/mL
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR