Humic acid

Supplier: BeanTown Chemical

130430-25G 130430-5G
BT130430-25GEA 169.18 USD
BT130430-25G BT130430-5G
Humic acid
Humic acid

CAS: 1415-93-6; EC No: 215-809-6; MDL No: MFCD00147177; RTECS: MT6544000 Crystalline/Powder


  • RTECS: MT6544000
  • Crystalline/Powder

Formula: C₉H₉NO₆
MDL Number: MFCD00147177
CAS Number: 1415-93-6
EINECS: 215-809-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR