Gold ≥99.95%, soluble APS 1,5-3,0 µm

Supplier: BeanTown Chemical

130420-1G 130420-5G
BT130420-1GEA 211 USD
BT130420-1G BT130420-5G
Gold ≥99.95%, soluble APS 1,5-3,0 µm
Gold

CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436 Solid; Molecular Formula: Au; MW: 196.97


  • Solid

Formula: Au
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR