1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

130395-10G 130395-250G 130395-50G
BT130395-10GEA 64.29 USD
BT130395-10G BT130395-250G BT130395-50G
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol

CAS: 647-42-7; EC No: 211-477-1; MDL No: MFCD00042143; RTECS: RH1460000 Liquid; Linear Formula: CF3(CF2)5(CH2)2OH; Molecular Formula: C8H5F13O; MW: 364.10 Boiling Point: 88-89°/28 mmHg; Flash point: 91°C (196°F) Density (g/mL): 1.651; Refractive Index: 1.313


  • RTECS: RH1460000
  • Refractive index: 1.313
  • Liquid

Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
Boiling Pt: 88…89 °C/28 mmHg
Density: 1.651 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR