Copper aluminum oxide ≥99.5%

Supplier: BeanTown Chemical

130375-250G 130375-50G
BT130375-250GEA 378 USD
BT130375-250G BT130375-50G
Copper aluminum oxide ≥99.5%
Copper aluminum oxide

CAS: 12042-92-1; EC No: 234-934-7; MDL No: MFCD00210610 -325 Mesh Powder; Molecular Formula: CuAl2O4; MW: 181.50


  • -325 Mesh Powder

Formula: CuAl₂O₄, may contain some CuO
MDL Number: MFCD00210610
CAS Number: 12042-92-1
EINECS: 234-934-7

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR