1,2-Dimethoxyethane ≥99% stabilised
Supplier: BeanTown Chemical
Synonyms:
Dimethyl glycol, Ethylene glycol dimethyl ether, DME
130215-100ML
130215-2.5L
130215-500ML
BT130215-100MLEA
44.36
USD
BT130215-100ML
BT130215-2.5L
BT130215-500ML
1,2-Dimethoxyethane ≥99% stabilised
1,2-Dimethoxyethane
CAS: 110-71-4; EC No: 203-794-9; MDL No: MFCD00008502; RTECS: KI1451000
UN No: UN2252; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3OCH2CH2OCH3; Molecular Formula: C4H10O2; MW: 90.12
Melting Point: -58°; Boiling Point: 85°; Flash point: -2°C (28°F)
Density (g/mL): 0.867; Refractive Index: 1.379
- RTECS: KI1451000
- Refractive index: 1.379
- Liquid
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 85 Melting Pt: -69 Density: 0.867 g/mL Flash Pt: –2 °C (28 °F) |
MDL Number:
MFCD00008502 CAS Number: 110-71-4 EINECS: 203-794-9 UN: 2252 ADR: 3 II Merck Index: 13,03251 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...