Ethyl acetate ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester
130040-1L
130040-4L
130040-500ML
BT130040-1LEA
40
USD
BT130040-1L
BT130040-4L
BT130040-500ML
Ethyl acetate ≥99.5% ACS
Ethyl acetate
CAS: 141-78-6; EC No: 205-500-4; MDL No: MFCD00009171; RTECS: AH5425000
UN No: UN1173; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COOCH2CH3; Molecular Formula: C4H8O2; MW: 88.11
Melting Point: -84°; Boiling Point: 76.5-77.5°; Flash point: -3°C (27°F)
Density (g/mL): 0.902; Refractive Index: 1.372
- RTECS: AH5425000
- Refractive index: 1.372
- Liquid
Formula:
CH₃COOC₂H₅ Boiling Pt: 76.5…77.5 °C Melting Pt: –84 °C Density: 0.902 g/mL Flash Pt: –3 °C (27 °F) |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% min. |
Specific gravity at 25°C | 0.894-0.898 |
Color (APHA) | ≤10 |
Residue after evaporation | ≤0.003% |
Titrable acid | ≤0.0009meq/g |
Substances darkened by H2SO4 | Passes test |
Limit of nonvolatile residue | ≤0.02% |
Acidity | ≤0.10ml of 0.10N NaOH for neutralization |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...