D-(+)-Cellobiose ≥98%

Supplier: BeanTown Chemical

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

130020-100G 130020-25G 130020-5G
BT130020-100GEA 206 USD
BT130020-100G BT130020-25G BT130020-5G
D-(+)-Cellobiose ≥98%
D(+)-Cellobiose

CAS: 528-50-7; EC No: 208-436-5; MDL No: MFCD00136034 Powder; Molecular Formula: C12H22O11; MW: 342.30 Melting Point: 239° (decomposes) Optical Rotation: [α]20/D +34±1°, c = 10% in water


  • Optical rotation: [α]20/D +34±1°, c = 10% in water
  • Powder

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 239 °C (dec.)
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR