Thiocarbohydrazide ≥97%
Supplier: BeanTown Chemical
Synonyms:
TCH
129755-100G
129755-25G
129755-5G
BT129755-100GEA
324
USD
BT129755-100G
BT129755-25G
BT129755-5G
Thiocarbohydrazide ≥97%
Thiocarbohydrazide
CAS: 2231-57-4; EC No: 218-769-8; MDL No: MFCD00007616; RTECS: FF2975000
UN No: UN2811; Haz Class: 6.1; Packing Group: I
Crystalline; Linear Formula: (NH2NH)2CS; Molecular Formula: CH6N4S; MW: 106.15
Melting Point: 171-174° (decomposes)
Light Sensitive
- RTECS: FF2975000
- Light Sensitive
- Crystalline
Formula:
CH₆N₄S Melting Pt: 171…174 °C (dec.) |
MDL Number:
MFCD00007616 CAS Number: 2231-57-4 EINECS: 218-769-8 UN: 2811 ADR: 6.1,I |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...