Nickel ≥99% (trace metals basis), foil, annealed, Thickness 0.1257 mm

Supplier: BeanTown Chemical

129715-20X30CM 129715-30X100 129715-30X500
BT129715-20X30EA 105.09 USD
BT129715-20X30 BT129715-30X10 BT129715-30X50
Nickel ≥99% (trace metals basis), foil, annealed, Thickness 0.1257 mm
Nickel

CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137; RTECS: VW4725000 Solid; Molecular Formula: Ni; MW: 58.69 Melting Point: 1453°; Boiling Point: 2732° Density (g/mL): 8.9


  • RTECS: VW4725000
  • Solid

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/mL
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR