1,3-Dithiane ≥98%
Supplier: BeanTown Chemical
129685-100G
129685-25G
129685-5G
BT129685-100GEA
553
USD
BT129685-100G
BT129685-25G
BT129685-5G
1,3-Dithiane ≥98%
1,3-Dithiane
CAS: 505-23-7; EC No: 208-006-7; MDL No: MFCD00006654; RTECS: JO5070000
UN No: UN3335; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C4H8S2; MW: 120.24
Melting Point: 52-54°; Flash point: 90°C (194°F)
- RTECS: JO5070000
- Solid
Formula:
C₄H₈S₂ Melting Pt: 52…54 °C Flash Pt: 90 °C (194 °F) |
MDL Number:
MFCD00006654 CAS Number: 505-23-7 EINECS: 208-006-7 UN: 3335 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...