Lithium cobaltite (lithium cobalt(III) oxide) ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Lithium cobalt(III) oxide

129515-100G 129515-25G
BT129515-100GEA 257 USD
BT129515-100G BT129515-25G
Lithium cobaltite (lithium cobalt(III) oxide) ≥99.5%
Lithium cobaltite (Lithium cobalt (III) oxide)

CAS: 12190-79-3; EC No: 235-362-0; MDL No: MFCD00049786 5 Micron APS Powder; Linear Formula: LiCoO2 ; MW: 97.88 Hygroscopic


  • Hygroscopic
  • 5 Micron APS Powder

Formula: CoLiO₂
MDL Number: MFCD00049786
CAS Number: 12190-79-3
EINECS: 235-362-0

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR