Styrene-4-sulfonic acid sodium salt

Supplier: BeanTown Chemical

Synonyms: 4-Vinylbenzenesulfonic acid sodium salt, Sodium 4-vinylbenzenesulphonate, p-Styrenesulfonic acid sodium salt, Sodium-p-styrenesulfonate, 4-Vinylbenzenesulphonic acid sodium salt, Sodium-p-styrenesulphonate, Sodium 4-vinylbenzenesulfonate, p-Styrenesulphonic acid sodium salt

129510-250G 129510-50G
BT129510-250GEA 132 USD
BT129510-250G BT129510-50G
Styrene-4-sulfonic acid sodium salt
Styrene-4-sulfonic acid sodium salt

CAS: 2695-37-6; EC No: 220-266-3; MDL No: MFCD00013379 Powder; Linear Formula: H2C=CHC6H4SO3Na; Molecular Formula: C8H7NaO3S; MW: 206.19 Melting Point: 300° Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Powder

Formula: C₈H₇NaO₃S
MW: 206.19 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00013379
CAS Number: 2695-37-6
EINECS: 220-266-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR