2-Chlorobenzylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: (2-Chlorophenyl)methanamine

129480-100G 129480-25G 129480-500G
BT129480-100GEA 176 USD
BT129480-100G BT129480-25G BT129480-500G
2-Chlorobenzylamine ≥98%
2-Chlorobenzylamine

CAS: 89-97-4; EC No: 203-245-3; MDL No: MFCD00008108 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H8ClN ; MW: 141.60 Boiling Point: 103-104°/11 mmHg; Flash point: 88°C (190°F) Density (g/mL): 1.173; Refractive Index: 1.562 Air Sensitive


  • Refractive index: 1.562
  • Air Sensitive
  • Liquid

Formula: C₇H₈ClN
Boiling Pt: 103…104 °C/11 mmHg
Density: 1.173 g/mL
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00008108
CAS Number: 89-97-4
EINECS: 203-245-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR