Propylene oxide ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide

129430-100ML 129430-1L
BT129430-100MLEA 30 USD
BT129430-100ML BT129430-1L
Propylene oxide ≥99%
Propylene oxide

CAS: 75-56-9; EC No: 200-879-2; MDL No: MFCD00005126; RTECS: TZ2975000 UN No: UN1280; Haz Class: 3; Packing Group: I Liquid; Molecular Formula: C3H6O; MW: 58.08 Melting Point: -112°; Boiling Point: 34-35°; Flash point: -37°C (-35°F) Density (g/mL): 0.83; Refractive Index: 1.366 Air Sensitive, Moisture Sensitive


  • RTECS: TZ2975000
  • Refractive index: 1.366
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34.3
Melting Pt: -112
Density: 0.83 g/mL
Flash Pt: –37 °C (–35 °F)
MDL Number: MFCD00005126
CAS Number: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3 I
Merck Index: 12,08041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR