3-(3-Methoxyphenyl)propionic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-(3-Methoxyphenyl)propanoic acid
129115-25G
129115-5G
BT129115-25GEA
160
USD
BT129115-25G
BT129115-5G
3-(3-Methoxyphenyl)propionic acid ≥98%
3-(3-Methoxyphenyl)propionic acid
CAS: 10516-71-9; EC No: 234-049-6; MDL No: MFCD00014027
Crystalline; Molecular Formula: C10H12O3; MW: 180.20
Melting Point: 43-45°
- Crystalline
Formula:
C₁₀H₁₂O₃ Melting Pt: 43…45 °C |
MDL Number:
MFCD00014027 CAS Number: 10516-71-9 EINECS: 234-049-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...