Cyclopropanemethylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: Cyclopropylmethylamine, (Cyclopropylmethyl)amine, (Aminomethyl)cyclopropane

129080-1G 129080-25G 129080-5G
BT129080-1GEA 49 USD
BT129080-1G BT129080-25G BT129080-5G
Cyclopropanemethylamine ≥98%
Cyclopropanemethylamine

CAS: 2516-47-4; EC No: 219-737-6; MDL No: MFCD00037147 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Molecular Formula: C4H9N; MW: 71.12 Boiling Point: 86°; Flash point: 2°C (36°F) Density (g/mL): 0.82


  • Liquid

Formula: C₄H₉N
Boiling Pt: 86 °C
Density: 0.82 g/mL
Flash Pt: 2 °C (36 °F)
MDL Number: MFCD00037147
CAS Number: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR