5-Chloro-2-pentanone ≥94%

Supplier: BeanTown Chemical

Synonyms: 5-Chloropentan-2-one

128960-100G 128960-25G 128960-500G
BT128960-100GEA 83 USD
BT128960-100G BT128960-25G BT128960-500G
5-Chloro-2-pentanone ≥94%
5-Chloro-2-pentanone

CAS: 5891-21-4; EC No: 227-565-8; MDL No: MFCD00001008; RTECS: SA8143500 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C5H9ClO ; MW: 120.58 Boiling Point: 71-72°/20 mm Hg; Flash point: 67°C (153°F) Density (g/mL): 1.057; Refractive Index: 1.438


  • RTECS: SA8143500
  • Refractive index: 1.438
  • Liquid

Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 71
Melting Pt: -48
Density: 1.057 g/mL
Flash Pt: 67 °C (153 °F)
MDL Number: MFCD00001008
CAS Number: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3 III

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR