Silicon nitride ≥95% 85% min α-phase

Supplier: BeanTown Chemical

Synonyms: Silicon(IV) nitride

128940-1KG 128940-250G 128940-50G
BT128940-1KGEA 527 USD
BT128940-1KG BT128940-250G BT128940-50G
Silicon nitride ≥95% 85% min α-phase
Silicon nitride

CAS: 12033-89-5; EC No: 234-796-8; MDL No: MFCD00011230; RTECS: VW0650000 1 Micron Powder, S.A. 5.0 m2/g; Linear Formula: Si3N4; MW: 140.28 Melting Point: 1900° (sublimes) Density (g/mL): 3.44


  • RTECS: VW0650000
  • <1 Micron Powder, S.A. 5.0 m²/g

Formula: N₄Si₃
Melting Pt: 1900 °C (subl.)
Density: 3.44 g/mL
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR