γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%
Supplier: BeanTown Chemical
128910-100G
128910-25G
128910-500G
BT128910-100GEA
90.04
USD
BT128910-100G
BT128910-25G
BT128910-500G
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99%
γ-(N-Morpholino)propanesulphonic acid (MOPS)
CAS: 1132-61-2; EC No: 214-478-5; MDL No: MFCD00006183; RTECS: QE9104530
Powder; Molecular Formula: C7H15NO4S; MW: 209.27
Melting Point: 277-280°; Flash point: 110°C (230°F)
- RTECS: QE9104530
- Powder
Formula:
C₇H₁₅NO₄S Melting Pt: 277…280 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00006183 CAS Number: 1132-61-2 EINECS: 214-478-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...