Tantalum ≥99.95% (trace metals basis), foil certified pinhole-free, Thickness 0.0127 mm

Supplier: BeanTown Chemical

128755-50X100 128755-50X2000 128755-50X500
BT128755-50X10EA 57 USD
BT128755-50X10 BT128755-50X20 BT128755-50X50
Tantalum ≥99.95% (trace metals basis), foil certified pinhole-free, Thickness 0.0127 mm
Tantalum

CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000 Solid; Molecular Formula: Ta; MW: 180.95 Melting Point: 2996°; Boiling Point: 5425° Density (g/mL): 16.69


  • RTECS: WW5505000
  • Solid

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/mL
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR