Dimethyl fumarate ≥99%

Supplier: BeanTown Chemical

Synonyms: Fumaric acid dimethyl ester

128695-100G 128695-250G 128695-25G
BT128695-100GEA 58 USD
BT128695-100G BT128695-250G BT128695-25G
Dimethyl fumarate ≥99%
Dimethyl fumarate

CAS: 624-49-7; EC No: 210-849-0; MDL No: MFCD00064438; RTECS: EM6125000 Powder; Linear Formula: CH3OCOCH=CHCOOCH3; Molecular Formula: C6H8O4 ; MW: 144.13 Melting Point: 102-106°; Boiling Point: 192-193°


  • RTECS: EM6125000
  • Powder

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192
Melting Pt: 102
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR