Germanium(IV) ethoxide ≥97%
Supplier: BeanTown Chemical
Synonyms:
Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate
128680-1G
128680-25G
128680-5G
BT128680-1GEA
52
USD
BT128680-1G
BT128680-25G
BT128680-5G
Germanium(IV) ethoxide ≥97%
Germanium (IV) ethoxide
CAS: 14165-55-0; EC No: 238-007-8; MDL No: MFCD00015121
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: Ge(OCH2CH3)4; Molecular Formula: C8H20GeO4 ; MW: 252.84
Flash point: 48°C (118°F)
Density (g/mL): 1.14 ; Refractive Index: 1.407
Moisture Sensitive
- Refractive index: 1.407
- Moisture Sensitive
- Liquid
Formula:
C₈H₂₀GeO₄ Density: 1.14 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00015121 CAS Number: 14165-55-0 EINECS: 238-007-8 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...