Acetophenone ≥99%

Supplier: BeanTown Chemical

128655-10L 128655-2.5L 128655-500ML
BT128655-10LEA 203 USD
BT128655-10L BT128655-2.5L BT128655-500ML
Acetophenone ≥99%
Acetophenone

CAS: 98-86-2; EC No: 202-708-7; MDL No: MFCD00008724; RTECS: AM5250000 Liquid; Molecular Formula: C8H8O; MW: 120.15 Melting Point: 19-20°; Boiling Point: 202°; Flash point: 76°C (169°F) Density (g/mL): 1.03; Refractive Index: 1.534


  • RTECS: AM5250000
  • Refractive index: 1.534
  • Liquid

Formula: C₈H₈O
Boiling Pt: 202 °C
Melting Pt: 19…20 °C
Density: 1.03 g/mL
Flash Pt: 76 °C (169 °F)
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR