Acetophenone ≥99%
Supplier: BeanTown Chemical
128655-10L
128655-2.5L
128655-500ML
BT128655-10LEA
203
USD
BT128655-10L
BT128655-2.5L
BT128655-500ML
Acetophenone ≥99%
Acetophenone
CAS: 98-86-2; EC No: 202-708-7; MDL No: MFCD00008724; RTECS: AM5250000
Liquid; Molecular Formula: C8H8O; MW: 120.15
Melting Point: 19-20°; Boiling Point: 202°; Flash point: 76°C (169°F)
Density (g/mL): 1.03; Refractive Index: 1.534
- RTECS: AM5250000
- Refractive index: 1.534
- Liquid
Formula:
C₈H₈O Boiling Pt: 202 °C Melting Pt: 19…20 °C Density: 1.03 g/mL Flash Pt: 76 °C (169 °F) |
MDL Number:
MFCD00008724 CAS Number: 98-86-2 EINECS: 202-708-7 UN: 3334 ADR: 9, |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...