Hexachloroplatinic(IV) acid hexahydrate ≥99.95% ACS

Supplier: BeanTown Chemical

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

128610-1G 128610-25G 128610-5G
BT128610-1GEA 148.36 USD
BT128610-1G BT128610-25G BT128610-5G
Hexachloroplatinic(IV) acid hexahydrate ≥99.95% ACS
Hexachloroplatinic (IV) acid hexahydrate

CAS: 18497-13-7; EC No: 241-010-7; MDL No: MFCD00149910; RTECS: TP1500000 UN No: UN2507; Haz Class: 8; Packing Group: III Lump; Linear Formula: H2PtCl6·6H2O; MW: 517.91 Melting Point: 60° (decomposes) Hygroscopic


  • RTECS: TP1500000
  • Hygroscopic
  • Lump

Formula: H₂PtCl₆·6H₂O
Melting Pt: 60 °C (dec.)
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III

Order Now

Specification Test Results

Assay 99.95%
Assay, Pt ≥37.50%
Solubility in alcohol Passes test
Alkali and other salts ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR