Terbium(III) chloride, anhydrous ≥99.99%

Supplier: BeanTown Chemical


Ampuled under argon

128525-10G 128525-1G
BT128525-10GEA 1049 USD
BT128525-10G BT128525-1G
Terbium(III) chloride, anhydrous ≥99.99%
Terbium (III) chloride

CAS: 10042-88-3; EC No: 233-132-4; MDL No: MFCD00011258 Powder; Linear Formula: TbCl3; MW: 265.29 Melting Point: 588° Density (g/mL): 4.35 Hygroscopic


  • Hygroscopic
  • Powder

Formula: Cl₃Tb
MW: 265.28
Melting Pt: 588
Density: 4.35 g/mL
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR