Aluminium ≥99.7%, soluble APS 11 µm
Supplier: BeanTown Chemical
Synonyms:
Aluminum
128475-1KG
128475-250G
128475-5KG
BT128475-1KGEA
152
USD
BT128475-1KG
BT128475-250G
BT128475-5KG
Aluminium ≥99.7%, soluble APS 11 µm
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000
UN No: UN3208; Haz Class: 4.3; Packing Group: II
Solid; Molecular Formula: Al; MW: 26.98
Melting Point: 660.4°; Boiling Point: 2460°
Density (g/mL): 2.7
- RTECS: BD0330000
- Solid
Formula:
Al Boiling Pt: 2460 °C Melting Pt: 660.4 °C Density: 2.7 g/mL |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 ADR: 4.3,II |
Specification Test Results
Assay | 99.7% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...