5-Acetyluracil ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Acetylpyrimidine-2,4(1H,3H)-dione
128375-1G
128375-25G
128375-5G
BT128375-1GEA
63
USD
BT128375-1G
BT128375-25G
BT128375-5G
5-Acetyluracil ≥98%
5-Acetyluracil
CAS: 6214-65-9; MDL No: MFCD00151968
Powder; Molecular Formula: C6H6N2O3; MW: 154.12
Melting Point: 278° (decomposes)
- Powder
Formula:
C₆H₆N₂O₃ Melting Pt: 278 °C (dec.) |
MDL Number:
MFCD00151968 CAS Number: 6214-65-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...