Cerium(IV) oxide ≥99.9%, pellets
Supplier: BeanTown Chemical
Synonyms:
Ceric oxide, Cerium dioxide
128360-100G
128360-500G
BT128360-100GEA
457
USD
BT128360-100G
BT128360-500G
Cerium(IV) oxide ≥99.9%, pellets
Cerium (IV) oxide
CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933; RTECS: FK6310000
Solid, approx. 9mm (0.35in) dia x 5mm (0.20in) thick; Molecular Formula: CeO2; MW: 172.12
Density (g/mL): 7.13
- RTECS: FK6310000
- Solid, approx. 9mm (0.35in) dia x 5mm (0.20in) thick
Formula:
CeO₂ Density: 7.13 g/mL |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...