Lithium nitrate hydrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical
Danger

128230-10G 128230-50G
BT128230-10GEA 61.78 USD
BT128230-10G BT128230-50G
Lithium nitrate hydrate ≥99.999% (trace metals basis)
Lithium nitrate hydrate

CAS: 7790-69-4; EC No: 232-218-9; MDL No: MFCD00011094 UN No: UN2722; Haz Class: 5.1; Packing Group: III Crystalline Aggregates; Molecular Formula: LiNO3·xH2O; MW: 68.95 (anhydrous) Hygroscopic


  • Crystalline Aggregates
  • Hygroscopic

Formula: LiNO₃·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 14,5536

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR