Aluminium ≥99% (trace metals basis), plate, Thickness 6.35 mm
Supplier: BeanTown Chemical
128185-25X25CM
BT128185-25X25EA
138
USD
BT128185-25X25
Aluminium ≥99% (trace metals basis), plate, Thickness 6.35 mm
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029
Solid; Molecular Formula: Al; MW: 26.98
Melting Point: 660.4°; Boiling Point: 2460°
Density (g/mL): 2.7
- Solid
Formula:
Al Boiling Pt: 2460 °C Melting Pt: 660.4 °C Density: 2.7 g/mL |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...