D-Tagatose ≥98%

Supplier: BeanTown Chemical

Synonyms: (3S,4S,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one

128160-1G 128160-250MG 128160-5G
BT128160-1GEA 117 USD
BT128160-1G BT128160-250MG BT128160-5G
D-Tagatose ≥98%
D-Tagatose

CAS: 87-81-0; EC No: 201-772-3; MDL No: MFCD00134449 Powder; Molecular Formula: C6H12O6; MW: 180.16


  • Powder

Formula: C₆H₁₂O₆
MDL Number: MFCD00134449
CAS Number: 87-81-0
EINECS: 201-772-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR