2,6-Dichloroaniline ≥98%
Supplier: BeanTown Chemical
|
Danger
|
128125-100G
128125-25G
BT128125-100GEA
83
USD
BT128125-100G
BT128125-25G
2,6-Dichloroaniline ≥98%
2,6-Dichloroaniline
CAS: 608-31-1; EC No: 210-160-5; MDL No: MFCD00007675; RTECS: CX9862655
UN No: UN3442; Haz Class: 6.1; Packing Group: II
Crystalline/Solid; Linear Formula: (Cl)2C6H3NH2; Molecular Formula: C6H5Cl2N; MW: 162.02
Melting Point: 37-40°; Boiling Point: 97°/5 mmHg; Flash point: 118°C (244°F)
Density (g/mL): 1.280
- RTECS: CX9862655
- Crystalline/Solid
Formula:
C₆H₅Cl₂N Boiling Pt: 97 °C (5 mmHg) Melting Pt: 37…40 °C Density: 1.280 (g/mL) Flash Pt: 118 °C |
MDL Number:
MFCD00007675 CAS Number: 608-31-1 EINECS: 210-160-5 UN: 3442 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...